Triethylammonium chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Triethylamine hydrochloride

222020-1KG 222020-250G
BT222020-1KGEA 121 USD
BT222020-1KG BT222020-250G
Triethylammonium chloride ≥98%
Triethylammonium chloride

CAS: 554-68-7; EC No: 209-067-2; MDL No: MFCD00012500; RTECS: YE2111500 Crystalline; Linear Formula: (CH3CH2)3N·HCl; Molecular Formula: C6H15N·HCl; MW: 137.65 Melting Point: 261° (decomposes) Density (g/mL): 1.07 Hygroscopic


  • RTECS: YE2111500
  • Hygroscopic
  • Crystalline

Formula: C₆H₁₆ClN
Melting Pt: 261 °C (dec.)
Density: 1.07 g/mL
MDL Number: MFCD00012500
CAS Number: 554-68-7
EINECS: 209-067-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR