Tantalum nitride ≥99.5%

Supplier: BeanTown Chemical

222005-10G 222005-50G
BT222005-10GEA 52 USD
BT222005-10G BT222005-50G
Tantalum nitride ≥99.5%
Tantalum nitride

CAS: 12033-62-4; EC No: 234-788-4; MDL No: MFCD00049568 -325 Mesh Powder; Linear Formula: TaN; MW: 194.95 Melting Point: 3360°


  • -325 Mesh Powder

Formula: NTa
MW: 194.95 g/mol
Melting Pt: 3360 °C
MDL Number: MFCD00049568
CAS Number: 12033-62-4
EINECS: 234-788-4

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR