Iron(III) oxide ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide

221995-100G 221995-25G 221995-500G
BT221995-100GEA 235 USD
BT221995-100G BT221995-25G BT221995-500G
Iron(III) oxide ≥99.99%
Iron(III) oxide

CAS: 1309-37-1; EC No: 215-168-2; MDL No: MFCD00011008 Powder; Molecular Formula: Fe2O3 ; MW: 159.69


  • Powder

Formula: Fe₂O₃
MW: 159.69 g/mol
Melting Pt: 1538
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR