2-(Boc-amino)-5-pyridinemethanol ≥97%

Supplier: BeanTown Chemical

Synonyms: tert-Butyl (5-(hydroxymethyl)pyridin-2-yl)carbamate

221970-1G 221970-5G
BT221970-1GEA 205 USD
BT221970-1G BT221970-5G
2-(Boc-amino)-5-pyridinemethanol ≥97%
2-(Boc-amino)-5-pyridinemethanol

CAS: 169280-83-5; MDL No: MFCD07186264 Solid; Molecular Formula: C11H16N2O3; MW: 224.26


  • Solid

Formula: C₁₁H₁₆N₂O₃
MW: 224.26 g/mol
MDL Number: MFCD07186264
CAS Number: 169280-83-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR