Niobium ≥99.99% (trace metals basis excluding Ta), wire Ø 0.25 mm
Supplier: BeanTown Chemical
221945-100CM
221945-25CM
221945-500CM
BT221945-100CMEA
62
USD
BT221945-100CM
BT221945-25CM
BT221945-500CM
Niobium ≥99.99% (trace metals basis excluding Ta), wire Ø 0.25 mm
Niobium
CAS: 7440-03-1; EC No: 231-113-5; MDL No: MFCD00011126
Powder; Molecular Formula: Nb ; MW: 92.91
Melting Point: 2468°; Boiling Point: 4742°
Density (g/mL): 8.57
- Powder
Formula:
Nb MW: 92.91 g/mol Boiling Pt: 4742 °C Melting Pt: 2468 °C Density: 8.57 g/mL |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...