2-Deoxy-D-glucose ≥98%
Supplier: BeanTown Chemical
Synonyms:
(3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal
221905-1G
221905-25G
221905-5G
BT221905-1GEA
63.43
USD
BT221905-1G
BT221905-25G
BT221905-5G
2-Deoxy-D-glucose ≥98%
2-Deoxy-D-glucose
CAS: 154-17-6; EC No: 205-823-0; MDL No: MFCD00151328; RTECS: MQ3325000
Crystalline/Powder; Molecular Formula: C6H12O5; MW: 164.16
Melting Point: 146-147°
- RTECS: MQ3325000
- Crystalline/Powder
Formula:
C₆H₁₂O₅ MW: 164.16 g/mol Melting Pt: 146…147 °C |
MDL Number:
MFCD00151328 CAS Number: 154-17-6 EINECS: 205-823-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...