6-Chloronicotinamide ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Chloropyridine-3-carboxamide

221835-1G 221835-25G 221835-5G
BT221835-1GEA 23 USD
BT221835-1G BT221835-25G BT221835-5G
6-Chloronicotinamide ≥98%
6-Chloronicotinamide

CAS: 6271-78-9; EC No: 228-456-8; MDL No: MFCD00006242 Powder; Molecular Formula: C6H5ClN2O; MW: 156.57 Melting Point: 210-212°


  • Powder

Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD00006242
CAS Number: 6271-78-9
EINECS: 228-456-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR