Tris(4-methylphenyl)phosphine ≥98%
Supplier: BeanTown Chemical
Synonyms:
Tri-p-tolylphosphine, Tris(p-tolyl)phosphine
221790-1G
221790-25G
221790-5G
BT221790-1GEA
18
USD
BT221790-1G
BT221790-25G
BT221790-5G
Tris(4-methylphenyl)phosphine ≥98%
Tris(4-methylphenyl)phosphine
CAS: 1038-95-5; EC No: 213-863-5; MDL No: MFCD00008542; RTECS: SZ3880000
Crystalline; Linear Formula: (CH3C6H4)3P; Molecular Formula: C21H21P; MW: 304.37
Melting Point: 144-148°
Air Sensitive
- RTECS: SZ3880000
- Air Sensitive
- Crystalline
Formula:
C₂₁H₂₁P MW: 304.37 g/mol Melting Pt: 144…148 °C |
MDL Number:
MFCD00008542 CAS Number: 1038-95-5 EINECS: 213-863-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...