Triethyl phosphonoacetate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate

221755-100G 221755-2.5KG 221755-500G
BT221755-100GEA 81 USD
BT221755-100G BT221755-2.5KG BT221755-500G
Triethyl phosphonoacetate ≥98%
Triethyl phosphonoacetate

CAS: 867-13-0; EC No: 212-757-6; MDL No: MFCD00009177; RTECS: AG9800000 Liquid; Linear Formula: (CH3CH2O)2P(O)CH2COOCH2CH3; Molecular Formula: C8H17O5P; MW: 224.19 Melting Point: -24°; Boiling Point: 260-262°; Flash point: 105°C (221°F) Density (g/mL): 1.125; Refractive Index: 1.431


  • RTECS: AG9800000
  • Refractive index: 1.431
  • Liquid

Formula: C₈H₁₇O₅P
Boiling Pt: 260…262 °C
Melting Pt: –24 °C
Density: 1.125 g/mL
Flash Pt: 105 °C (221 °F)
MDL Number: MFCD00009177
CAS Number: 867-13-0
EINECS: 212-757-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR