Molybdenum ≥99.95% (trace metals basis), slug, Length 12.7 mm, Diameter 6.35 mm
Supplier: BeanTown Chemical
221750-100G
221750-25G
BT221750-100GEA
133
USD
BT221750-100G
BT221750-25G
Molybdenum ≥99.95% (trace metals basis), slug, Length 12.7 mm, Diameter 6.35 mm
Molybdenum
CAS: 7439-98-7; EC No: 231-107-2; MDL No: MFCD00003465
Solid; Molecular Formula: Mo; MW: 95.94
Melting Point: 2617°; Boiling Point: 4612°
Density (g/mL): 10.3
- Solid
Formula:
Mo Boiling Pt: 4612 °C Melting Pt: 2617 °C Density: 10.3 g/mL |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...