Bismuth(III) citrate ≥94%
Supplier: BeanTown Chemical
221735-1KG
221735-250G
BT221735-1KGEA
299
USD
BT221735-1KG
BT221735-250G
Bismuth(III) citrate ≥94%
Bismuth(III) citrate
CAS: 813-93-4; EC No: 212-390-1; MDL No: MFCD00050817
Powder, typically 10 micron max ; Molecular Formula: C6H5BiO7; MW: 398.08
Melting Point: 300° (decomposes)
Density (g/mL): 0.94
Light Sensitive
- Light Sensitive
- Powder, typically 10 micron max
Formula:
C₆H₅BiO₇ Melting Pt: 300 °C (dec.) Density: 0.94 g/mL |
MDL Number:
MFCD00050817 CAS Number: 813-93-4 EINECS: 212-390-1 |
Specification Test Results
Assay | 94% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...