(±)-2-Ethyl-1-hexanol ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Ethylhexan-1-ol
221730-2.5L
221730-500ML
BT221730-2.5LEA
77.38
USD
BT221730-2.5L
BT221730-500ML
(±)-2-Ethyl-1-hexanol ≥99%
(±)-2-Ethyl-1-hexanol
CAS: 104-76-7; EC No: 203-234-3; MDL No: MFCD00004746; RTECS: MP0350000
Liquid; Molecular Formula: C8H18O; MW: 130.23
Melting Point: -76°; Boiling Point: 183-186°; Flash point: 81°C (178°F)
Density (g/mL): 0.833; Refractive Index: 1.431
- RTECS: MP0350000
- Refractive index: 1.431
- Liquid
Formula:
CH₃(CH₂)₃CH(C₂H₅)CH₂OH MW: 130.23 g/mol Boiling Pt: 183…186 °C Melting Pt: –76 °C Density: 0.833 g/mL Flash Pt: 81 °C (178 °F) |
MDL Number:
MFCD00004746 CAS Number: 104-76-7 EINECS: 203-234-3 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...