Dilithium zirconium trioxide ≥94%
Supplier: BeanTown Chemical
Synonyms:
Lithium zirconate, Lithium zirconium oxide
221725-100G
221725-25G
221725-500G
BT221725-100GEA
248.43
USD
BT221725-100G
BT221725-25G
BT221725-500G
Dilithium zirconium trioxide ≥94%
Dilithium zirconium trioxide
CAS: 12031-83-3; EC No: 234-760-1; MDL No: MFCD00016182
-80 Mesh Powder; Linear Formula: Li2ZrO3; MW: 153.1
- -80 Mesh Powder
Formula:
Li₂O₃Zr MW: 153.1 |
MDL Number:
MFCD00016182 CAS Number: 12031-83-3 EINECS: 234-760-1 |
Specification Test Results
Assay | 94% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...