4-Phenoxybenzoic acid ≥99%
Supplier: BeanTown Chemical
221695-25G
221695-5G
BT221695-25GEA
128
USD
BT221695-25G
BT221695-5G
4-Phenoxybenzoic acid ≥99%
4-Phenoxybenzoic acid
CAS: 2215-77-2; EC No: 218-682-5; MDL No: MFCD00002539
Solid; Molecular Formula: C13H10O3; MW: 214.22
Melting Point: 163-165°
- Solid
Formula:
C₁₃H₁₀O₃ MW: 214.22 g/mol Melting Pt: 163…165 °C |
MDL Number:
MFCD00002539 CAS Number: 2215-77-2 EINECS: 218-682-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...