Methyl-3,4-diaminobenzoate ≥98%
Supplier: BeanTown Chemical
Synonyms:
3,4-Diamino benzoate, 3,4-Diaminobenzoic acid methyl ester, Methyl 3,4-diaminobenzoate
221680-100G
221680-25G
221680-5G
BT221680-100GEA
1118
USD
BT221680-100G
BT221680-25G
BT221680-5G
Methyl-3,4-diaminobenzoate ≥98%
Methyl-3,4-diaminobenzoate
CAS: 36692-49-6; EC No: 000-000-0; MDL No: MFCD00017098
Solid; Molecular Formula: C8H10N2O2; MW: 166.18
Melting Point: 105-110°
- Solid
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol Melting Pt: 105…110 °C |
MDL Number:
MFCD00017098 CAS Number: 36692-49-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...