2-Fluorobenzonitrile ≥99%
Supplier: BeanTown Chemical
221645-100G
221645-10G
221645-50G
BT221645-100GEA
262
USD
BT221645-100G
BT221645-10G
BT221645-50G
2-Fluorobenzonitrile ≥99%
2-Fluorobenzonitrile
CAS: 394-47-8; MDL No: MFCD00001773
Liquid; Molecular Formula: C7H4FN ; MW: 121.11
Boiling Point: 90°/21 mmHg; Flash point: 74°C (165°F)
Density (g/mL): 1.116; Refractive Index: 1.505
- Refractive index: 1.505
- Liquid
Formula:
C₇H₄FN Boiling Pt: 90 °C/21 mmHg Density: 1.116 g/mL Flash Pt: 74 °C (165 °F) |
MDL Number:
MFCD00001773 CAS Number: 394-47-8 UN: 3276 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...