1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Methoxy-4-[(4-pentylphenyl)ethynyl]benzene, 1-(4-Methoxyphenyl)-2-(4-pentylphenyl)acetylene, 1-(4-Methoxyphenyl)ethynyl-4-pentylbenzene

221640-1G 221640-5G
BT221640-1GEA 58 USD
BT221640-1G BT221640-5G
1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene ≥99%
1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene

CAS: 39969-28-3; MDL No: MFCD00187208 Crystalline/Powder; Molecular Formula: C20H22O; MW: 278.39 Melting Point: 47°; Boiling Point: 168-170°/0.5 mmHg


  • Crystalline/Powder

Formula: C₂₀H₂₂O
MW: 278.395 g/mol
Boiling Pt: 168…170 °C/0.5 mmHg
Melting Pt: 47 °C
MDL Number: MFCD00187208
CAS Number: 39969-28-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR