Zirconium carbonate basic hydrate

Supplier: BeanTown Chemical

221625-2KG 221625-500G
BT221625-2KGEA 136 USD
BT221625-2KG BT221625-500G
Zirconium carbonate basic hydrate
Zirconium carbonate basic hydrate

CAS: 57219-64-4; EC No: 260-633-5; MDL No: MFCD00172265 Powder; Linear Formula: (ZrO2)2·CO2·xH2O; MW: 290.45 (anhydrous) Density (g/mL): 2.2


  • Powder

Formula: C₄H₄O₁₂Zr
MW: 335.288 g/mol
Density: 2.2 g/mL
MDL Number: MFCD00172265
CAS Number: 57219-64-4
EINECS: 260-633-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR