4-Bromoanisole ≥99%
Supplier: BeanTown Chemical
CAS: 104-92-7; EC No: 203-252-1; MDL No: MFCD00000097; RTECS: BZ8501000
Liquid; Molecular Formula: C7H7BrO; MW: 187.03
Melting Point: 9-10°; Boiling Point: 223°; Flash point: 98°C (208°F)
Density (g/mL): 1.494; Refractive Index: 1.564
- RTECS: BZ8501000
- Refractive index: 1.564
- Liquid
Formula:
C₇H₇BrO Boiling Pt: 223 °C Melting Pt: 9…10 °C Density: 1.494 g/mL Flash Pt: 98 °C (208 °F) |
MDL Number:
MFCD00000097 CAS Number: 104-92-7 EINECS: 203-252-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...