Ammonium carbonate ≥99.999%

Supplier: BeanTown Chemical

Synonyms: Salt of hartshorn

221610-10G 221610-250G 221610-50G
BT221610-10GEA 40 USD
BT221610-10G BT221610-250G BT221610-50G
Ammonium carbonate ≥99.999%
Ammonium carbonate

CAS: 506-87-6; EC No: 208-058-0; MDL No: MFCD00010890; RTECS: BP1925000 Powder; Linear Formula: (NH4)2CO3; MW: 96.09 Hygroscopic


  • RTECS: BP1925000
  • Hygroscopic
  • Powder

Formula: (NH₄)₂CO₃
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR