3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid ≥98%

Supplier: BeanTown Chemical

221600-25G 221600-5G
BT221600-25GEA 353 USD
BT221600-25G BT221600-5G
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid ≥98%
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid

CAS: 20170-32-5; EC No: 243-556-1; MDL No: MFCD00017519 Powder; Molecular Formula: C17H26O3; MW: 278.40 Melting Point: 172-175°


  • Powder

Formula: C₁₇H₂₆O₃
Melting Pt: 172…175 °C
MDL Number: MFCD00017519
CAS Number: 20170-32-5
EINECS: 243-556-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR