Silicone oil Oilbath, temperature_high
Supplier: BeanTown Chemical
Synonyms:
Polyphenyl-methylsiloxane
221590-100G
221590-2.5KG
221590-500G
BT221590-100GEA
78.82
USD
BT221590-100G
BT221590-2.5KG
BT221590-500G
Silicone oil Oilbath, temperature_high
Silicone oil
Silicone oil, for oil baths, 25 to 250°C (open system) and 25 to 315°C (closed system)
- RTECS: VW3146875
- Refractive Index: 1.505
Density:
1.07 g/mL Flash Pt: >300 °C (>572 °F) |
MDL Number:
MFCD00132673 CAS Number: 68083-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...