4-Chloro-4'-hydroxybenzophenone ≥97%
Supplier: BeanTown Chemical
Synonyms:
(4-Chlorophenyl)(4-hydroxyphenyl)methanone
221575-10G
221575-250G
221575-50G
BT221575-10GEA
44
USD
BT221575-10G
BT221575-250G
BT221575-50G
4-Chloro-4'-hydroxybenzophenone ≥97%
4-Chloro-4'-hydroxybenzophenone
CAS: 42019-78-3; EC No: 255-627-4; MDL No: MFCD00002357
Liquid; Molecular Formula: C13H9ClO2 ; MW: 232.66
Melting Point: 175-178°; Boiling Point: 257°/13 mmHg
- Liquid
Formula:
C₁₃H₉ClO₂ MW: 232.67 g/mol Boiling Pt: 257 °C/13 mmHg Melting Pt: 175…178 °C |
MDL Number:
MFCD00002357 CAS Number: 42019-78-3 EINECS: 255-627-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...