Iridium ≥99.95% (trace metals basis), Powder -60 mesh
Supplier: BeanTown Chemical
221560-10G
221560-2G
221560-500MG
BT221560-10GEA
4144
USD
BT221560-10G
BT221560-2G
BT221560-500MG
Iridium ≥99.95% (trace metals basis), Powder -60 mesh
Iridium
CAS: 7439-88-5; EC No: 231-095-9; MDL No: MFCD00011062
Powder; Molecular Formula: Ir; MW: 192.22
Melting Point: 2450°; Boiling Point: 4130°
Density (g/mL): 22.65
- Powder
Formula:
Ir MW: 192.22 g/mol Boiling Pt: 4130 °C Melting Pt: 2450 °C Density: 22.65 g/mL |
MDL Number:
MFCD00011062 CAS Number: 7439-88-5 EINECS: 231-095-9 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...