1,3-Propanediol ≥99%

Supplier: BeanTown Chemical

221500-1KG 221500-250G 221500-5KG
BT221500-1KGEA 245.31 USD
BT221500-1KG BT221500-250G BT221500-5KG
1,3-Propanediol ≥99%
1,3-Propanediol

CAS: 504-63-2; EC No: 207-997-3; MDL No: MFCD00002949; RTECS: TY2010000 Liquid; Linear Formula: HO(CH2)3OH; Molecular Formula: C3H8O2; MW: 76.09 Melting Point: -27°; Boiling Point: 214°; Flash point: 110°C (230°F) Density (g/mL): 1.053; Refractive Index: 1.440


  • RTECS: TY2010000
  • Refractive index: 1.440
  • Liquid

Formula: HO(CH₂)₃OH
Boiling Pt: 214 °C
Melting Pt: –27 °C
Density: 1.053 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00002949
CAS Number: 504-63-2
EINECS: 207-997-3

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR