1,4-Butanediol ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,4-Dihydroxy butane, BDO, Tetramethylene glycol, 1,4-Butylene glycol

221435-1KG 221435-2.5KG 221435-250G
BT221435-1KGEA 45.52 USD
BT221435-1KG BT221435-2.5KG BT221435-250G
1,4-Butanediol ≥99%
1,4-Butanediol

CAS: 110-63-4; EC No: 203-786-5; MDL No: MFCD00002968; RTECS: EK0525000 Liquid; Linear Formula: HO(CH2)4OH; Molecular Formula: C4H10O2; MW: 90.12 Melting Point: 16°; Boiling Point: 230°; Flash point: 135°C (275°F) Density (g/mL): 1.017; Refractive Index: 1.445 Hygroscopic


  • RTECS: EK0525000
  • Refractive index: 1.445
  • Hygroscopic
  • Liquid

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 230 °C
Melting Pt: 16 °C
Density: 1.017 g/mL
Flash Pt: 135 °C (275 °F)
MDL Number: MFCD00002968
CAS Number: 110-63-4
EINECS: 203-786-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR