Chromium lanthanum trioxide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Lanthanum chromite
221400-10G
221400-50G
BT221400-10GEA
71
USD
BT221400-10G
BT221400-50G
Chromium lanthanum trioxide ≥99.5%
Chromium lanthanum trioxide
CAS: 12017-94-6; EC No: 234-629-9; MDL No: MFCD00053990
-200 Mesh Powder; Linear Formula: LaCrO3; MW: 238.90
- -200 Mesh Powder
Formula:
CrLaO₃ MW: 238.9 |
MDL Number:
MFCD00053990 CAS Number: 12017-94-6 EINECS: 234-629-9 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...