β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98%

Supplier: BeanTown Chemical

Synonyms: Tetra-O-acetyl-β-D-ribofuranose, (2S,3R,4R,5R)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate

221395-100G 221395-25G 221395-5G
BT221395-100GEA 506 USD
BT221395-100G BT221395-25G BT221395-5G
β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98%
beta-D-Ribofuranose-1,2,3,5-tetraacetate

CAS: 13035-61-5; EC No: 235-898-5; MDL No: MFCD00005358 Crystalline; Molecular Formula: C13H18O9; MW: 318.28 Melting Point: 81-83° Optical Rotation: [α]26/D −11.4°, c = 10 in chloroform


  • Optical rotation: [α]26/D −11.4°, c = 10 in chloroform
  • Crystalline

Formula: C₁₃H₁₈O₉
Melting Pt: 81…83 °C
MDL Number: MFCD00005358
CAS Number: 13035-61-5
EINECS: 235-898-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR