Magnesium stearate

Supplier: BeanTown Chemical

221355-1KG 221355-250G
BT221355-1KGEA 145 USD
BT221355-1KG BT221355-250G
Magnesium stearate
Magnesium stearate
Formula: C₃₆H₇₀MgO₄
MDL Number: MFCD00036391
CAS Number: 557-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR