Salicylaldehyde ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Hydroxybenzaldehyde
221335-100G
221335-2.5KG
221335-500G
BT221335-100GEA
34.61
USD
BT221335-100G
BT221335-2.5KG
BT221335-500G
Salicylaldehyde ≥99%
Salicylaldehyde
CAS: 90-02-8; EC No: 201-961-0; MDL No: MFCD00003317; RTECS: VN5250000
Liquid; Linear Formula: HOC6H4CHO; Molecular Formula: C7H6O2; MW: 122.12
Melting Point: -7°; Boiling Point: 197°; Flash point: 77°C (171°F)
Density (g/mL): 1.146; Refractive Index: 1.573
Air Sensitive, Light Sensitive
- RTECS: VN5250000
- Refractive index: 1.573
- Air Sensitive, Light Sensitive
- Liquid
Formula:
C₇H₆O₂ MW: 122.12 g/mol Boiling Pt: 197 °C Melting Pt: –7 °C Density: 1.146 g/mL Flash Pt: 77 °C (171 °F) |
MDL Number:
MFCD00003317 CAS Number: 90-02-8 EINECS: 201-961-0 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...