Phloroglucinol dihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,3,5-Trihydroxybenzene dihydrate, Phloroglucinol, Hydrate

221310-100G 221310-25G 221310-500G
BT221310-100GEA 84.95 USD
BT221310-100G BT221310-25G BT221310-500G
Phloroglucinol dihydrate ≥98%
Phloroglucinol dihydrate

CAS: 6099-90-7; EC No: 203-611-2; MDL No: MFCD00149090; RTECS: DC4662500 Powder; Linear Formula: C6H3(OH)3·2H2O; MW: 162.14 Melting Point: 218-221°; Flash point: 100°C (212°F) Light Sensitive


  • RTECS: DC4662500
  • Light Sensitive
  • Powder

Formula: C₆H₆O₃·2H₂O
MW: 162.14 g/mol
Melting Pt: 218…221 °C
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00149090
CAS Number: 6099-90-7
EINECS: 203-611-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR