1-Nonanesulfonic acid sodium salt ≥98%

Supplier: BeanTown Chemical

221290-25G 221290-5G
BT221290-25GEA 231 USD
BT221290-25G BT221290-5G
1-Nonanesulfonic acid sodium salt ≥98%
1-Nonanesulphonic acid sodium salt

CAS: 35192-74-6; MDL No: MFCD00025010 Powder; Linear Formula: CH3(CH2)8SO3Na; Molecular Formula: C9H19NaO3S; MW: 230.30 Melting Point: 300°


  • Powder

Formula: C₉H₁₉NaO₃S
Melting Pt: >300 °C
MDL Number: MFCD00025010
CAS Number: 35192-74-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR