3-Decanol ≥97%
Supplier: BeanTown Chemical
221245-100G
221245-25G
221245-5G
BT221245-100GEA
606
USD
BT221245-100G
BT221245-25G
BT221245-5G
3-Decanol ≥97%
3-Decanol
CAS: 1565-81-7; MDL No: MFCD00021956
Liquid; Molecular Formula: C10H22O; MW: 158.29
Boiling Point: 213°; Flash point: 99°C (210°F)
Density (g/mL): 0.830
- Liquid
Formula:
C₁₀H₂₂O MW: 158.29 g/mol Boiling Pt: 213 °C Density: 0.830 g/mL Flash Pt: 99 °C (210 °F) |
MDL Number:
MFCD00021956 CAS Number: 1565-81-7 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...