Pyrene ≥98%

Supplier: BeanTown Chemical

221205-100G 221205-25G
BT221205-100GEA 95 USD
BT221205-100G BT221205-25G
Pyrene ≥98%
Pyrene

CAS: 129-00-0; EC No: 204-927-3; MDL No: MFCD00004136; RTECS: UR2450000 Crystalline; Molecular Formula: C16H10; MW: 202.26 Melting Point: 145-148°; Boiling Point: 390-395°; Flash point: 208°C (406°F) Density (g/mL): 1.271


  • RTECS: UR2450000
  • Crystalline

Formula: C₁₆H₁₀
MW: 202.26 g/mol
Boiling Pt: 390…395 °C
Melting Pt: 145…148 °C
Density: 1.271 g/mL
Flash Pt: 208 °C (406 °F)
MDL Number: MFCD00004136
CAS Number: 129-00-0
EINECS: 204-927-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b03eb6b87b60315a282988993f066fd2d3af129&vid=d3f1ffa103d650d2e9f49139db04842dfa6ca0ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR