N-Boc-4-oxo-L-proline ≥98%

Supplier: BeanTown Chemical

Synonyms: Boc-4-oxo-Pro-OH, (S)-1-Boc-4-oxo-2-pyrrolidinecarboxylic Acid, (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, (S)-1-(tert-Butoxycarbonyl)-4-oxo-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4-oxo-L-proline

221185-1G 221185-250MG 221185-5G
BT221185-1GEA 203 USD
BT221185-1G BT221185-250MG BT221185-5G
N-Boc-4-oxo-L-proline ≥98%
N-Boc-4-oxo-L-proline

CAS: 84348-37-8; MDL No: MFCD01860669 Solid; Molecular Formula: C10H15NO5; MW: 229.24 Melting Point: 160° (decomposes) Optical Rotation: [α]22/D +19.0 to +23.0°, c = 0.5 in acetone


  • Optical rotation: [α]22/D +19.0 to +23.0°, c = 0.5 in acetone
  • Solid

Formula: C₁₀H₁₅NO₅
Melting Pt: 160 °C (dec.)
MDL Number: MFCD01860669
CAS Number: 84348-37-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR