L(-)-Menthol ≥99%
Supplier: BeanTown Chemical
Synonyms:
(1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol
221175-1KG
221175-250G
221175-50G
BT221175-1KGEA
261
USD
BT221175-1KG
BT221175-250G
BT221175-50G
L(-)-Menthol ≥99%
L(-)-Menthol
CAS: 2216-51-5; EC No: 218-690-9; MDL No: MFCD00062979; RTECS: OT0700000
Crystalline; Molecular Formula: C10H20O; MW: 156.27
Melting Point: 41-45°; Boiling Point: 212°; Flash point: 101°C (214°F)
Optical Rotation: [α]20/D -50°, c = 10 in ethanol
- RTECS: OT0700000
- Optical rotation: [α]20/D -50°, c = 10 in ethanol
- Crystalline
Formula:
C₁₀H₂₀O MW: 156.27 g/mol Boiling Pt: 212 °C Melting Pt: 41…45 °C Flash Pt: 101 °C (214 °F) |
MDL Number:
MFCD00062979 CAS Number: 2216-51-5 EINECS: 218-690-9 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...