4-Phenyl-1-butyne ≥98%
Supplier: BeanTown Chemical
221170-25G
221170-5G
BT221170-25GEA
341
USD
BT221170-25G
BT221170-5G
4-Phenyl-1-butyne ≥98%
4-Phenyl-1-butyne
CAS: 16520-62-0; MDL No: MFCD00041667
Liquid; Molecular Formula: C10H10; MW: 130.19
Boiling Point: 189-190°
Density (g/mL): 0.926; Refractive Index: 1.512
- Refractive index: 1.512
- Liquid
Formula:
C₁₀H₁₀ Boiling Pt: 189…190 °C Density: 0.926 g/mL |
MDL Number:
MFCD00041667 CAS Number: 16520-62-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...