Lanthanum hexaboride ≥99.5% (trace rare earth metals basis)

Supplier: BeanTown Chemical

Synonyms: Lanthanum boride

221160-10G 221160-250G 221160-50G
BT221160-10GEA 71.38 USD
BT221160-10G BT221160-250G BT221160-50G
Lanthanum hexaboride ≥99.5% (trace rare earth metals basis)
Lanthanum hexaboride

CAS: 12008-21-8; EC No: 234-531-6; MDL No: MFCD00151350 -325 Mesh Powder; Linear Formula: LaB6; MW: 203.78 Melting Point: 2210° Density (g/mL): 2.61


  • -325 Mesh Powder

Formula: B₆La
MW: 203.77 g/mol
Melting Pt: 2210 °C
Density: 2.61 g/mL
MDL Number: MFCD00151350
CAS Number: 12008-21-8
EINECS: 234-531-6

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR