Dipotassium hexachloropalladate

Supplier: BeanTown Chemical

Synonyms: Potassium hexachloropalladate(IV)

221135-1G 221135-5G
BT221135-1GEA 46.47 USD
BT221135-1G BT221135-5G
Dipotassium hexachloropalladate
Dipotassium hexachloropalladate

CAS: 16919-73-6; EC No: 240-974-6; MDL No: MFCD00043081; RTECS: RT3353000 Crystalline/Powder; Linear Formula: K2PdCl6; MW: 397.32 Density (g/mL): 2.738


  • RTECS: RT3353000
  • Crystalline/Powder

Formula: Cl₆K₂Pd
MW: 397.33 g/mol
Density: 2.738 g/mL
MDL Number: MFCD00043081
CAS Number: 16919-73-6
EINECS: 240-974-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR