Indium (III) oxide ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Indium trioxide, Diindium trioxide

221130-10G 221130-250G 221130-50G
BT221130-10GEA 82 USD
BT221130-10G BT221130-250G BT221130-50G
Indium (III) oxide ≥99.995%
Indium (III) oxide

CAS: 1312-43-2; EC No: 215-193-9; MDL No: MFCD00011060 Powder; Molecular Formula: In2O3; MW: 277.64 Density (g/mL): 7.18


  • Powder

Formula: In₂O₃
MW: 277.64 g/mol
Melting Pt: 715.05
Density: 7.18 g/mL
MDL Number: MFCD00011060
CAS Number: 1312-43-2
EINECS: 215-193-9
Merck Index: 12,04984

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR