5-Methyl-1H-tetrazole ≥98%

Supplier: BeanTown Chemical

221110-25G 221110-5G
BT221110-25GEA 155 USD
BT221110-25G BT221110-5G
5-Methyl-1H-tetrazole ≥98%
5-Methyl-1H-tetrazole

CAS: 4076-36-2; MDL No: MFCD00129971 Crystalline; Molecular Formula: C2H4N4; MW: 84.08 Melting Point: 142-146°


  • Crystalline

Formula: C₂H₄N₄
MW: 84.08 g/mol
Melting Pt: 142…146 °C
MDL Number: MFCD00185824
CAS Number: 4076-36-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR