2-Bromo-3,3,3-trifluoropropene ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Bromo-3,3,3-trifluoro-1-propene

221070-25G 221070-5G
BT221070-25GEA 215.02 USD
BT221070-25G BT221070-5G
2-Bromo-3,3,3-trifluoropropene ≥97%
2-Bromo-3,3,3-trifluoropropene

CAS: 1514-82-5; MDL No: MFCD00077469 Liquid; Molecular Formula: C3H2BrF3; MW: 174.95 Boiling Point: 29-30° Refractive Index: 1.353


  • Refractive index: 1.353
  • Liquid

Formula: C₃H₂BrF₃
MW: 174.95 g/mol
Boiling Pt: 29…30 °C
MDL Number: MFCD00077469
CAS Number: 1514-82-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR