Chlorhexidine gluconate 20% (w/w) in water

Supplier: BeanTown Chemical

Synonyms: 1,1'-Hexamethylenebis[5-(4-chlorophenyl)biguanide] digluconate, Peridex, 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide] (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid(1:2), Chlorhexidine digluconate

221065-100ML 221065-25ML 221065-500ML
BT221065-100MLEA 71.55 USD
BT221065-100ML BT221065-25ML BT221065-500ML
Chlorhexidine gluconate 20% (w/w) in water
Chlorhexidine gluconate

CAS: 18472-51-0; EC No: 242-354-0; MDL No: MFCD00083599 Liquid; Molecular Formula: C22H30Cl2N10·2C6H12O7 ; MW: 897.76 Density (g/mL): 1.06 Light Sensitive


  • Light Sensitive
  • Liquid

Formula: C₂₂H₃₀Cl₂N₁₀·2C₆H₁₂O₇
MW: 897.76 g/mol
Density: 1.06 g/mL
MDL Number: MFCD00083599
CAS Number: 18472-51-0
EINECS: 242-354-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR