Potassium sulfate ≥99.0% ACS
Supplier: BeanTown Chemical
221055-2KG
221055-500G
BT221055-2KGEA
189.53
USD
BT221055-2KG
BT221055-500G
Potassium sulfate ≥99.0% ACS
Potassium sulphate
CAS: 7778-80-5; EC No: 231-915-5; MDL No: MFCD00011388; RTECS: TT5900000
Crystalline; Linear Formula: K2SO4; MW: 174.26
Melting Point: 1069°; Boiling Point: 1689°
Density (g/mL): 2.662
- RTECS: TT5900000
- Crystalline
Formula:
K₂SO₄ Boiling Pt: 1689 °C Melting Pt: 1069 °C Density: 2.662 |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 EINECS: 231-915-5 Merck Index: 14,7674 |
Specification Test Results
Assay | 99.0% min. |
Insoluble matter | ≤0.01% |
Calcium (Ca) | ≤0.01% |
Magnesium (Mg) | ≤0.005% |
Sodium (Na) | ≤0.02% |
Chloride (Cl) | ≤0.001% |
Nitrogen compounds (as N) | ≤5ppm |
Heavy metals (as Pb) | ≤5ppm |
Iron (Fe) | ≤5ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...