2-(2-(Piperazin-1-yl)ethoxy)ethanol ≥95%

Supplier: BeanTown Chemical

Synonyms: 1-[2-(2-Hydroxyethoxy)ethyl]piperazine, 1-Hydroxyethylethoxypiperazine

221035-1G 221035-25G 221035-5G
BT221035-1GEA 13 USD
BT221035-1G BT221035-25G BT221035-5G
2-(2-(Piperazin-1-yl)ethoxy)ethanol ≥95%
2-(2-(Piperazin-1-yl)ethoxy)ethanol

CAS: 13349-82-1; MDL No: MFCD00074931 Liquid; Molecular Formula: C8H18N2O2 ; MW: 174.24 Boiling Point: 112-114°/0.25 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.061; Refractive Index: 1.497


  • Refractive index: 1.497
  • Liquid

Formula: C₈H₁₈N₂O₂
MW: 174.2407 g/mol
Boiling Pt: 112…114 °C/0.25 mmHg
Density: 1.061 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00074931
CAS Number: 13349-82-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR