Cbz-1-amino-1-cyclopentanecarboxylic acid

Supplier: BeanTown Chemical

Synonyms: 1-(Cbz-amino)cyclopentanecarboxylic Acid

220975-1G 220975-5G
BT220975-1GEA 133 USD
BT220975-1G BT220975-5G
Cbz-1-amino-1-cyclopentanecarboxylic acid
Cbz-1-amino-1-cyclopentanecarboxylic acid

CAS: 17191-44-5; MDL No: MFCD02094399 Solid; Molecular Formula: C14H17NO4; MW: 263.29 Boiling Point: 468°


  • Solid

Formula: C₁₄H₁₇NO₄
MW: 263.293 g/mol
Boiling Pt: 468 °C
MDL Number: MFCD02094399
CAS Number: 17191-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR