2-Methyl-6-quinolinol ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Hydroxy-2-methylquinoline

220940-1G 220940-25G 220940-5G
BT220940-1GEA 85 USD
BT220940-1G BT220940-25G BT220940-5G
2-Methyl-6-quinolinol ≥98%
2-Methyl-6-quinolinol

CAS: 613-21-8; MDL No: MFCD00047600 Needles; Molecular Formula: C10H9NO; MW: 159.19


  • Needles

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD00047600
CAS Number: 613-21-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR