Adenine hydrochloride hydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: 6-Aminopurine hydrochloride hydrate

220920-100G 220920-25G 220920-5G
BT220920-100GEA 447 USD
BT220920-100G BT220920-25G BT220920-5G
Adenine hydrochloride hydrate ≥99%
Adenine hydrochloride hydrate

CAS: 6055-72-7; MDL No: MFCD00150864 Powder; Molecular Formula: C5H5N5·HCl·xH2O; MW: 171.59 (anhydrous) Melting Point: 289-291° (decomposes)

Formula: C₅H₅N₅·HCl·xH₂O
MW: 171.59 g/mol (anhy)
Melting Pt: 289…291 °C (dec.)
MDL Number: MFCD00150864
CAS Number: 916986-40-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR