Bis(diphenylphosphino)methane ≥97%

Supplier: BeanTown Chemical

Synonyms: [(1,1-Diphenylphosphino)methyl](diphenyl)phosphine, DPPM

220915-25G 220915-5G
BT220915-25GEA 178 USD
BT220915-25G BT220915-5G
Bis(diphenylphosphino)methane ≥97%
Bis(diphenylphosphino)methane

CAS: 2071-20-7; EC No: 218-194-2; MDL No: MFCD00003537 Crystalline; Linear Formula: (C6H5)2PCH2P(C6H5)2; Molecular Formula: C25H22P2; MW: 384.39 Melting Point: 118-119° Air Sensitive


  • Air Sensitive
  • Crystalline

Formula: C₂₅H₂₂P₂
Melting Pt: 118…119 °C
MDL Number: MFCD00003537
CAS Number: 2071-20-7
EINECS: 218-194-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR