Holmium(III) oxide ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Diholmium trioxide, Holmium oxide
220910-10G
220910-50G
BT220910-10GEA
124
USD
BT220910-10G
BT220910-50G
Holmium(III) oxide ≥99.99%
Holmium(III) oxide
CAS: 12055-62-8; EC No: 235-015-3; MDL No: MFCD00011053
Powder; Molecular Formula: Ho2O3; MW: 377.86
Melting Point: 2330°
- Powder
Formula:
Ho₂O₃ MW: 377.86 g/mol Boiling Pt: 2600 Melting Pt: 2400 |
MDL Number:
MFCD00011053 CAS Number: 12055-62-8 EINECS: 235-015-3 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...